Learn More
Invitrogen™ FKBP25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593277
Description
Immunogen sequence: MAAAVPQRAW TVEQLRSEQL PKKDIIKFLQ EHGSDSFLAE HKLLGNIKNV AKTANKDHLV TAYNHLFETK RFKGTESISK VSEQVKNVKL NEDKPKETKS EETLDEGPPK YTKSVLKKGD KTNFPKKGDV VHCWYTGTLQ DGTVFDTNIQ TSAKKKKNAK PLSFKVGVGK VIRGWDEALL TMSKGEKARL EIEPEWAYGK KGQPDAKIPP NAKLTFEVEL VDID; Positive Samples: HeLa, MCF-7, BT-474; Cellular Location: Nucleus.
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.
Specifications
FKBP25 | |
Polyclonal | |
Unconjugated | |
FKBP3 | |
25 kDa FK506-binding protein; 25 kDa FKBP; 25kDa; FK506 binding protein 3; FK506 binding protein 3, 25kDa; FK506-binding protein 25, T-cell; FK506-binding protein 3; FK506-binding protein 3 (25kD); FKBP25; FKBP-25; Fkbp3; FKBP-3; Immunophilin FKBP25; peptidyl-prolyl cis-trans isomerase FKBP3; PPIase; PPIase FKBP3; rapamycin binding protein; rapamycin-selective 25 kDa immunophilin; Rotamase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2287 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q00688 | |
FKBP3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-224 of human FKBP3 (NP_0020041). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.