Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519483
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519483 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519483 Supplier Invitrogen™ Supplier No. PA519483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with rabbit and cow based on sequence homology.

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP25
Applications Western Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene FKBP3
Gene Accession No. Q00688, Q62446
Gene Alias 25 kDa FK506-binding protein; 25 kDa FKBP; 25kDa; FK506 binding protein 3; FK506 binding protein 3, 25kDa; FK506-binding protein 25, T-cell; FK506-binding protein 3; FK506-binding protein 3 (25kD); FKBP25; FKBP-25; Fkbp3; FKBP-3; Immunophilin FKBP25; peptidyl-prolyl cis-trans isomerase FKBP3; PPIase; PPIase FKBP3; rapamycin binding protein; rapamycin-selective 25 kDa immunophilin; Rotamase
Gene Symbols FKBP3
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human FKBP25.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2287, 299104, 30795
Target Species Human, Mouse, Rat, Non-human Primate, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.