Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115238
Description
Antibody detects endogenous levels of total FKBP25.
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.
Specifications
| FKBP3 | |
| Polyclonal | |
| Unconjugated | |
| FKBP3 | |
| 25 kDa FK506-binding protein; 25 kDa FKBP; 25kDa; FK506 binding protein 3; FK506 binding protein 3, 25kDa; FK506-binding protein 25, T-cell; FK506-binding protein 3; FK506-binding protein 3 (25kD); FKBP25; FKBP-25; Fkbp3; FKBP-3; Immunophilin FKBP25; peptidyl-prolyl cis-trans isomerase FKBP3; PPIase; PPIase FKBP3; rapamycin binding protein; rapamycin-selective 25 kDa immunophilin; Rotamase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2287, 299104, 30795 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q00688, Q62446 | |
| FKBP3 | |
| A synthesized peptide derived from human FKBP3(Accession Q00688), corresponding to amino acid residues E97-T147. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction