Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP38/FKBP8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982678
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2678 Supplier Bioss Supplier No. BS13179R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.

Specifications

Antigen FKBP38/FKBP8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FKBP8
Gene Accession No. Q14318
Gene Alias 38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; BOS_7129; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; Fkbp38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; Peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; Rotamase; Sam11
Gene Symbols FKBP8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FKBP38/FKBP8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23770
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.