Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP38 Rabbit anti-Human, Mouse, Rat, Clone: 2J7P6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | FKBP38 |
---|---|
Clone | 2J7P6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
FKBP38 Monoclonal antibody specifically detects FKBP38 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FKBP38 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Apoptosis, Cancer, Cellular Markers, Core ESC Like Genes, Glia Markers, Growth and Development, Mitotic Regulators, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23770 | |
IgG | |
Affinity purified |
2J7P6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FKBP38 (Q14318). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title