Learn More
FKBP4/FKBP52 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | FKBP4/FKBP52 |
|---|---|
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-0361
|
Bethyl Laboratories, Inc
A301426AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It has high structural and functional similarity to FK506-binding protein 1A and thus may play a role in the intracellular trafficking of hetero-oligomeric forms of the steroid hormone receptors. This protein correlates strongly with adeno-associated virus type 2 vectors resulting in a significant increase in AAV-mediated transgene expression in human cell lines. Thus this encoded protein is thought to have important implications for the optimal use of AAV vectors in human gene therapy. The human genome contains several non-transcribed pseudogenes similar to this gene.Specifications
| FKBP4/FKBP52 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| FKBP51; FKBP59; HBI; Hsp56; PPIase; p52; 51 kDa FK506-binding protein; 59 kDa FK506-binding protein 4; HSP binding immunophilin; T-cell FK506-binding protein; peptidyl-prolyl cis-trans isomerase FKBP4; peptidylprolyl cis-trans isomerase; rotamase | |
| Between 1 and 50 | |
| Primary | |
| 4° C | |
| FKBP4 |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P30416, Q02790 | |
| 14228, 2288 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.