Learn More
FKBP5/FKBP51 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301430AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.Specifications
| FKBP5/FKBP51 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q13451 | |
| FKBP5 | |
| Between 407 and 457 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| FKBP5 | |
| FK506 Binding Protein 51, EC 5.2.1.8, Peptidyl-prolyl cis-trans isomerase, PPIase, Rotamase, 51 kDa FK506-binding protein, 54 kDa progesterone recepto | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2289 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.