Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKBP5 Monoclonal Antibody (Hi51B), PerCP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545162
This item is not returnable.
View return policy
Description
A 1:2,000 dilution was sufficient for detection of FKBP51 in approximately 50 μg total protein using WB analysis. Detects approximately 51kDa.
FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.
Specifications
FKBP5 | |
Monoclonal | |
1 mg/mL | |
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative | |
Q13451, Q64378 | |
FKBP5 | |
Synthetic peptide corresponding to the residues of human FKBP51. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit, Hamster | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
Hi51B | |
PerCP | |
FKBP5 | |
51 kDa FK506-binding protein; 51 kDa FKBP; 54 kDa progesterone receptor-associated immunophilin; AIG6; androgen-regulated protein 6; D17Ertd592e; Dit1; EC 5.2.1.8; FF1 antigen; FK506 binding protein 5; FK506-binding protein 5; FKBP prolyl isomerase 5; Fkbp5; FKBP-5; FKBP51; FKBP-51; FKBP54; HSP90-binding immunophilin; p54; peptidylprolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase FKBP5; PPIase; PPIase FKBP5; Ptg-10; rotamase; T-cell FK506-binding protein | |
Mouse | |
Protein G | |
RUO | |
100339155, 100760904, 14229, 2289, 361810, 481759 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction