Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP5 Polyclonal Antibody

Catalog No. PIPA5121984
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121984 20 μL
PIPA5119119 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121984 Supplier Invitrogen™ Supplier No. PA5121984
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FKBP5 Polyclonal Antibody for Western Blot, ICC/IF, IHC

FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP5
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FKBP5
Gene Accession No. Q13451, Q64378
Gene Alias 51 kDa FK506-binding protein; 51 kDa FKBP; 54 kDa progesterone receptor-associated immunophilin; AIG6; androgen-regulated protein 6; D17Ertd592e; Dit1; EC 5.2.1.8; FF1 antigen; FK506 binding protein 5; FK506-binding protein 5; FKBP prolyl isomerase 5; Fkbp5; FKBP-5; FKBP51; FKBP-51; FKBP54; HSP90-binding immunophilin; p54; peptidylprolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase FKBP5; PPIase; PPIase FKBP5; Ptg-10; rotamase; T-cell FK506-binding protein
Gene Symbols FKBP5
Host Species Rabbit
Immunogen Recombinant protein FK506 Binding Protein 5. The antigen corresponds to amino acid range 2-457 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14229, 2289
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.