Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP6 Antibody (9B7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242645AF532

Catalog No. NP24264A32


Only null left
Add to Cart

Description

Description

FKBP6 Monoclonal specifically detects FKBP6 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

FKBP6
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
FKBP6
Recombinant human FKBP6 (1-327aa) purified from E. coli
0.1 mL
8468.0
Store at 4°C in the dark.
Western Blot, ELISA
9B7
Western Blot, ELISA
36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase
Mouse
Protein A purified
Primary
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.