Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP6 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297729R
Description
FKBP6 Polyclonal specifically detects FKBP6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FKBP6 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase | |
Produced in rabbits immunized with E. coli-derived Human FKBP6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8468 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction