Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145151 Supplier Invitrogen™ Supplier No. PA5145151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEEQHLVEAA KLPVLLNLSF TYLKLDRPTI ALCYGEQALI IDQKNAKALF RCGQACLLLT EYQKARDFLV RAQKEQPFNH DINNELKKLA S Antigen sequence identity: Mouse 84%; Rat 84%.

FKBP6 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP6 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin.This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fkbp6
Gene Accession No. O75344
Gene Alias 1700008G22Rik; 36 kDa FK506 binding protein; 36 kDa FK506-binding protein; 36 kDa FKBP; 36kDa; As; AU017274; D5Ertd724e; FK506 binding protein 6; FK506 binding protein 6, 36kDa; FK506-binding protein 6; FK506-binding protein 6 (36kD); FKBP prolyl isomerase 6; FKBP prolyl isomerase family member 6 (inactive); Fkbp36; FKBP-36; FKBP6; FKBP-6; Immunophilin FKBP36; Inactive peptidyl-prolyl cis-trans isomerase FKBP6; Inactive PPIase FKBP6; MGC87179; peptidyl-prolyl cis-trans isomerase FKBP6; PPiase; PPIase FKBP6; rotamase
Gene Symbols Fkbp6
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 208-298.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8468
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.