Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP7 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299915JF549
Description
FKBP7 Polyclonal antibody specifically detects FKBP7 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
FKBP7 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
EC 5.2.1.8, FK506 binding protein 7, FK506-binding protein 7FK506-binding protein 23,23 kDa FK506-binding protein, FKBP-23, FKBP23PPIase, FKBP-7, MGC9420, peptidyl-prolyl cis-trans isomerase FKBP7,23 kDa FKBP, PPIase FKBP7, rotamase | |
Produced in rabbits immunized with purified, recombinant Human FKBP7 (Uniprot#: Q9Y680-2; Met1-Gln218) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51661 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction