Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP7 Polyclonal Antibody

Catalog No. PIPA5121983
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121983 20 μL
PIPA5119120 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121983 Supplier Invitrogen™ Supplier No. PA5121983
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FKBP7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family. Members of this family exhibit PPIase activity and function as molecular chaperones. A similar protein in mouse is located in the endoplasmic reticulum and binds calcium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FKBP7
Gene Accession No. Q9Y680
Gene Alias 23 kDa FK506-binding protein; 23 kDa FKBP; 23kDa; FK506 binding protein 7; FK506-binding protein 23; FK506-binding protein 7; FKBP23; FKBP-23; Fkbp7; FKBP-7; Peptidyl-prolyl cis-trans isomerase FKBP7; PPIase; PPIase FKBP7; rotamase; UNQ670/PRO1304
Gene Symbols FKBP7
Host Species Rabbit
Immunogen Recombinant protein FK506 Binding Protein 7. The antigen corresponds to amino acid range 24-217 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.