Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547513
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547513 Supplier Invitrogen™ Supplier No. PA547513
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, less than 1% cross-reactivity with recombinant human FKBP-12, -12.6, -13, -25, -51, or -52 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene FKBP8
Gene Accession No. O35465, Q14318, Q3B7U9
Gene Alias 38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; FKBP38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; rotamase; Sam11
Gene Symbols FKBP8
Host Species Goat
Immunogen E. coli-derived recombinant mouse FKBP38 Met1-Gly326.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14232, 23770, 290652
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.