Learn More
Invitrogen™ FKBP8 Polyclonal Antibody

Description
Immunogen sequence: QPPAEEAEQP GALAREFLAA MEPEPAPAPA PEEWLDILGN GLLRKKTLVP GPPGSSRPVK GQVVTVHLQT SLENGTRVQE EPELVFTLGD CDVIQALDLS VPLMDVGETA MVTADSKYCY GPQGSRSPYI PPHAALCLEV TLKTAVDGPD LEMLTGQERV ALANRKRECG NAHYQRADFV LAANSYDLAI KAITSSAKVD MTFEEEAQLL Q; Positive Samples: Jurkat, THP-1, BT-474, HeLa, HL-60, Mouse liver, Mouse brain, Mouse eye, Rat brain; Cellular Location: Cytoplasmic side, Mitochondrion membrane, Single-pass membrane protein.
Specifications
Specifications
| Antigen | FKBP8 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FKBP8 |
| Gene Accession No. | O35465, Q14318, Q3B7U9 |
| Gene Alias | 38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; FKBP38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; rotamase; Sam11 |
| Gene Symbols | FKBP8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.