Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA583763
Description
Immunogen sequence: YDLAIKAITS SAKVDMTFEE EAQLLQLKVK CLNNLAASQL KLDHYRAALR SCSLVLEHQP DNIKALFRKG KVLAQQGEYS EAIPILRAAL KLEPSNKTIH AELSKLVKKH AAQRSTETAL YRKMLGNPSR LPAKCPGK
FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.
Specifications
FKBP8 | |
Polyclonal | |
Unconjugated | |
FKBP8 | |
38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; FKBP38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; rotamase; Sam11 | |
Rabbit | |
Anitgen affinity chromatography, Antigen affinity chromatography | |
RUO | |
23770 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14318 | |
FKBP8 | |
Recombinant protein corresponding to Human FKBP8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction