Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP8 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990863
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0863 25 μL
50-199-0862 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0863 Supplier Rockland Immunochemicals Supplier No. 200401A58S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human FKBP8 protein. A BLAST analysis was used to suggest partial cross-reactivity with FKBP8 from mouse and rat sources based on ∼93% homology with the immunizing sequence. Cross-reactivity with FKBP8 from other sources has not been determined.

FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.

Specifications

Antigen FKBP8
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene FKBP8
Gene Accession No. O35465, Q14318, Q3B7U9
Gene Alias 38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; BOS_7129; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; Fkbp38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; Peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; Rotamase; Sam11
Gene Symbols FKBP8
Host Species Rabbit
Immunogen This protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human FKBP8 protein.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14232, 23770, 290652
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.