Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514280
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514280 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514280 Supplier Invitrogen™ Supplier No. PA514280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

FKBP9 is a PPIase which accelerate the folding of proteins during protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP9
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FKBP9
Gene Accession No. O95302, Q9Z247
Gene Alias 1810014L23; 63 kDa FK506-binding protein; 63 kDa FKBP; AW549641; FK506 binding protein 9; FK506 binding protein 9, 63 kDa; FK506-binding protein 9; FKBP60; FKBP63; FKBP-63; FKBP65RS; Fkbp9; FKBP-9; Peptidyl-prolyl cis-trans isomerase FKBP9; PPIase; PPIase FKBP9; rotamase
Gene Symbols FKBP9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 541-570 amino acids from the C-terminal region of human FKBP9.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11328, 27055
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.