Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552952 Supplier Invitrogen™ Supplier No. PA552952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGMDQALVGM CVNERRFVKI PPKLAYGNEG VSGVIPPNSV LHFDVLLMDI WNSEDQVQIH TYFKPPSCPR TIQVSDF Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

FKBP9 is a PPIase which accelerate the folding of proteins during protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FKBP9
Gene Accession No. O95302
Gene Alias 1810014L23; 63 kDa FK506-binding protein; 63 kDa FKBP; AW549641; FK506 binding protein 9; FK506 binding protein 9, 63 kDa; FK506-binding protein 9; FKBP60; FKBP63; FKBP-63; FKBP65RS; Fkbp9; FKBP-9; Peptidyl-prolyl cis-trans isomerase FKBP9; PPIase; PPIase FKBP9; rotamase
Gene Symbols FKBP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FKBP9. Recombinant protein control fragment (Product #RP-90742).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.