Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBPL Antibody (OTI2D8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272125F
Description
FKBPL Monoclonal specifically detects FKBPL in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
FKBPL | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FKBPL | |
Full length recombinant protein of human FKBPL (NP_071393) produced in HEK293T cells. | |
0.1 mL | |
63943.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2D8 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DIR1WISP39, FK506 binding protein like, FK506-binding protein like, NG7FK506-binding protein-like, WAF-1/CIP1 stabilizing protein 39, WISp39 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction