Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBPL Monoclonal Antibody (OTI2D8)
GREENER_CHOICE

Catalog No. PIMA525356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525356 Supplier Invitrogen™ Supplier No. MA525356
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FKBPL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBPL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2D8
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fkbpl
Gene Accession No. Q9UIM3
Gene Alias DIR1; FK506 binding protein like; FK506 binding protein-like; FK506-binding protein like; FK506-binding protein-like; FKBP4; FKBPL; immunophilin-like protein NG7; NG7; peptidyl-prolyl cis-trans isomerase; Ppiase-X; WAF-1/CIP1 stabilizing protein 39; WISp39
Gene Symbols Fkbpl
Host Species Mouse
Immunogen Full length human recombinant protein of FKBPL produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63943
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.