Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563857 Supplier Invitrogen™ Supplier No. PA563857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETARYVVGVL EAAGVRYWLE GGSLLGAARH GDIIPWDYDV DLGIYLEDVG NCEQLRGAEA GSVVDERGFV WEKAVEGDF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a protein which is targeted to the medial Golgi apparatus and is necessary for posttranslational modification of dystroglycan. Mutations in this gene have been associated with congenital muscular dystrophy, mental retardation, and cerebellar cysts. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq, Oct 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKRP
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FKRP
Gene Accession No. Q9H9S5
Gene Alias A830029B19Rik; AI842067; AI847300; FKRP; fukutin related protein; fukutin-related protein; LGMD1I; LGMD2I; MDC1C; MDDGA5; MDDGB5; MDDGC5; Ribitol-5-phosphate transferase
Gene Symbols FKRP
Host Species Rabbit
Immunogen Recombinant protein fragment of Human FKRP (Product #RP-99940)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79147
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.