Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKRP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565349
Description
Immunogen sequence: ETARYVVGVLE AAGVRYWLEG GSLLGAARHG DIIPWDYDVD LGIYLEDVGN CEQLRGAEAG SVVDERGFVW EKAVEGDF Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This gene encodes a protein which is targeted to the medial Golgi apparatus and is necessary for posttranslational modification of dystroglycan. Mutations in this gene have been associated with congenital muscular dystrophy, mental retardation, and cerebellar cysts. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq, Oct 2008].
Specifications
FKRP | |
Polyclonal | |
Unconjugated | |
FKRP | |
A830029B19Rik; AI842067; AI847300; FKRP; fukutin related protein; fukutin-related protein; LGMD1I; LGMD2I; MDC1C; MDDGA5; MDDGB5; MDDGC5; Ribitol-5-phosphate transferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79147 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H9S5 | |
FKRP | |
Recombinant Human FKRP. Recombinant protein control fragment (Product #RP-99940). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction