Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKSG2 Polyclonal Antibody

FKSG2 Polyclonal Antibody
Supplier: Thermo Scientific PA575596
Description
FKSG2 Polyclonal Antibody for Western Blot

Specifications
FKSG2 | |
Polyclonal | |
Unconjugated | |
TPT1P8 | |
FKSG2; TPT1P8; tumor protein, translationally-controlled 1 pseudogene 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59347 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAU6 | |
TPT1P8 | |
Synthetic peptide corresponding to residues in Human FKSG2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction