Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKTN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542692 Supplier Invitrogen™ Supplier No. PA542692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TAFALQYHLW KNEEGWFRIA ENMGFQCLKI ESKDPRLDGI DSLSGTEIPL Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%.

FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKTN
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FKTN
Gene Accession No. O75072
Gene Alias CMD1X; D830030O17Rik; Fcmd; FKTN; Fukutin; Fukuyama type congenital muscular dystrophy (fukutin); Fukuyama type congenital muscular dystrophy homolog; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; LGMD2M; LOW QUALITY PROTEIN: fukutin; MDDGA4; MDDGB4; MDDGC4; patient fukutin; Ribitol-5-phosphate transferase; RP11-235C23.1
Gene Symbols FKTN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FKTN (aa 111-160).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2218
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.