Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKTN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100533 Supplier Invitrogen™ Supplier No. PA5100533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Fukutin.

FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKTN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FKTN
Gene Accession No. O75072, Q8R507
Gene Alias CMD1X; D830030O17Rik; Fcmd; FKTN; Fukutin; Fukuyama type congenital muscular dystrophy (fukutin); Fukuyama type congenital muscular dystrophy homolog; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; LGMD2M; LOW QUALITY PROTEIN: fukutin; MDDGA4; MDDGB4; MDDGC4; patient fukutin; Ribitol-5-phosphate transferase; RP11-235C23.1
Gene Symbols FKTN
Host Species Rabbit
Immunogen A synthesized peptide derived from human FKTN(Accession O75072), corresponding to amino acid residues E124-V174.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2218, 246179, 362520
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.