Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKTN Recombinant Rabbit Monoclonal Antibody (7J5N6)
GREENER_CHOICE

Catalog No. PIMA542665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542665 Supplier Invitrogen™ Supplier No. MA542665
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: BxPC-3, RD, U-87MG, Mouse testis, Mouse brain, Mouse heart, Rat liver. The target is usually found in the following locations: Golgi apparatus membrane, Single-pass type II membrane protein. Immunogen sequence: DVKLDVFFFY EETDHMWNGG TQAKTGKKFK YLFPKFTLCW TEFVDMKVHV PCETLEYIEA NYGKTWKIPV KTWDWKRSPP NVQPNGIWPI SEWDEVIQLY.

FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKTN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7J5N6
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FKTN
Gene Accession No. O75072, Q8R507
Gene Alias CMD1X; D830030O17Rik; Fcmd; FKTN; Fukutin; Fukuyama type congenital muscular dystrophy (fukutin); Fukuyama type congenital muscular dystrophy homolog; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; LGMD2M; LOW QUALITY PROTEIN: fukutin; MDDGA4; MDDGB4; MDDGC4; patient fukutin; Ribitol-5-phosphate transferase; RP11-235C23.1
Gene Symbols FKTN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 362-461 of human Fukutin (O75072).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2218, 246179, 362520
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.