Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543310 Supplier Invitrogen™ Supplier No. PA543310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGRSVTAGII IVGDEILKGH TQDTNTFFLC RTLRSLGVQV CRVSVVPDEV Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLAD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FLAD1
Gene Accession No. Q8NFF5
Gene Alias A930017E24Rik; fa92c06; FAD pyrophosphorylase; FAD synthase; FAD synthase region; FAD synthetase; FAD1; FAD1 flavin adenine dinucleotide synthetase homolog; Fad1, flavin adenine dinucleotide synthetase, homolog; FADS; FAD-synthetase; FLAD1; Flavin adenine dinucleotide synthase; flavin adenine dinucleotide synthetase; flavin adenine dinucleotide synthetase 1; FMN adenylyltransferase; hypothetical protein LOC506682; LSMFLAD; Molybdenum cofactor biosynthesis protein-like region; PP591; RFad1, flavin adenine dinucleotide synthetase, homolog (yeast); RP11-307C12.7; wu:fa92c06; zgc:91843
Gene Symbols FLAD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FLAD1 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80308
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.