Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLAD1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FLAD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FLAD1 Polyclonal specifically detects FLAD1 in Human, Mouse samples. It is validated for Western Blot.Specifications
FLAD1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
80308 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
EC 2.7.7, FAD1, FAD1 flavin adenine dinucleotide synthetase homolog (S. cerevisiae), Fad1, flavin adenine dinucleotide synthetase, homolog (yeast), FAD-synthetase, Flavin adenine dinucleotide synthase, flavin adenine dinucleotide synthetase, flavin adenine dinucleotide synthetase, homolog, MGC31803, MGC40255, PP591 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 388-587 of human FLAD1 (NP_079483.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title