Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLAG Epitope (DYKDDDDK) Chicken anti-Chemical, Polyclonal, Gallus

Chicken Polyclonal Antibody
Supplier: Vector Laboratories AFLAG

This item is not returnable.
View return policy
Description
FLAG Epitope (DYKDDDDK) Polyclonal antibody specifically detects FLAG Epitope (DYKDDDDK) in Chemical samples. It is validated for ELISA, Immunohistochemistry, Western BlotSpecifications
FLAG Epitope (DYKDDDDK) | |
Polyclonal | |
Unconjugated | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
FLAG Epitope (DYKDDDDK) | |
0.1 mg | |
Primary | |
Antibody | |
IgY |
ELISA, Immunohistochemistry, Western Blot | |
0.5-2 mg/mL | |
PBS with 0.1% sodium azide; pH | |
Chicken | |
Affinity chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction