Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLAG peptide, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against recombinant FLAG peptide.
Supplier: Abnova Corporation PAB29056
Description
Sequence: DYKDDDDKSpecifications
| FLAG peptide | |
| Polyclonal | |
| Chicken polyclonal antibody raised against recombinant FLAG peptide. | |
| In PBS, pH 7.2 (0.02% sodium azide) | |
| A KLH-conjugated synthetic peptide corresponding to Flag tag emulsified in Freund's adjuvant. | |
| 1 mL | |
| Primary | |
| Liquid |
| ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA Immunocytochemistry(1:500) Immunohistochemistry Immunoprecipitation(1:500) Western Blot(1:1000) The optimal working dilution should be determined by the end user. | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| Store at 4°C and avoid from light. For long term storage dilute the antibody with sterile glycerol and store it at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction