Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flagellin Antibody (06), Alexa Fluor™ 750 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305967AF750
Description
Flagellin Monoclonal antibody specifically detects Flagellin in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Flagellin | |
Monoclonal | |
Alexa Fluor 750 | |
Mouse | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
Western Blot, ELISA | |
06 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Listeria monocytogenes Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction