Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flagellin Antibody (121), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290335AF700
Description
Flagellin Monoclonal specifically detects Flagellin in Bacteria samples. It is validated for ELISA.Specifications
Flagellin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
This antibody was obtained from a rabbit immunized with purified, recombinant Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287). | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
121 | |
ELISA | |
Rabbit | |
Protein A purified | |
RUO | |
Listeria monocytogenes Listeria monocytogenes flagellin/FlaA | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction