Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flagellin Antibody (121), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290335MFV450
Description
Flagellin Monoclonal antibody specifically detects Flagellin in Bacteria samples. It is validated for ELISASpecifications
Flagellin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit immunized with purified, recombinant Listeria monocytogenes Flagellin (Accession#: YP_013330.1, Met1-Ser287) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
ELISA | |
121 | |
ELISA | |
Rabbit | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction