Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flagellin Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306390AF594
This item is not returnable.
View return policy
Description
Flagellin Polyclonal antibody specifically detects Flagellin in Bacteria samples. It is validated for ELISA, ImmunofluorescenceSpecifications
| Flagellin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| Bacteria | |
| Purified |
| ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction