Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flagellin Rabbit anti-Bacteria, Clone: 121, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290335100UL
Description
Flagellin Monoclonal specifically detects Flagellin in Bacteria samples. It is validated for ELISA.Specifications
Flagellin | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Recombinant Listeria Flagellin protein | |
100 μg | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
121 | |
Flow Cytometry, ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
Rabbit | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction