Learn More
Fisherbrand™ Flange Plug Caps, 13 mm
Catalog No. 02681512 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Green
Lavender
Natural
Orange
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-681-512 Black
02-681-509 Blue
02-681-510 Natural
02-681-511 Green
02-681-513 Lavender
02-681-514 Orange
02-681-515 Red
02-681-516 White
02-681-517 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 02-681-512
Add to Cart
Add to Cart
This item is not returnable. View return policy
These polyethylene (PE) flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes and round cuvettes.
  • Two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps.
Specifications
Closure Size 13 mm
Autoclavable Not Autoclavable
Material Polyethylene
Color Black
Quantity 1000/Cs.