Learn More
Fisherbrand™ Flange Plug Caps, 16 mm
Catalog No. 02681520 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Green
Lavender
Natural
Orange
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-681-520 Green
02-681-518 Blue
02-681-519 Natural
02-681-521 Black
02-681-522 Lavender
02-681-523 Orange
02-681-524 Red
02-681-525 White
02-681-526 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 02-681-520
Add to Cart
Add to Cart
This item is not returnable. View return policy
These polyethylene (PE) flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes and round cuvettes.
  • Two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps.
Specifications
Closure Size 16 mm
Autoclavable Not Autoclavable
Material Polyethylene
Color Green
Quantity 1000/Cs.
Sterility Non-sterile
Disposable Single-use