Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLAP Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299845PCP
Description
FLAP Polyclonal antibody specifically detects FLAP in Human samples. It is validated for ELISASpecifications
FLAP | |
Polyclonal | |
ELISA | |
arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein | |
Produced in rabbits immunized with purified, recombinant Human FLAP (Accession#: NP_001620.2; Met1-Pro161) | |
0.1 mL | |
Immunology, Inflammation | |
241 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction