Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLAP Antibody, R&D Systems™
SDP

Catalog No. NB462856 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB462856 100 μg
NB462855 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB462856 Supplier R&D Systems Supplier No. NBP330926100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

FLAP Polyclonal specifically detects FLAP in Human samples. It is validated for Western Blot.

Specifications

Antigen FLAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1-5 μg/mL
Formulation Phosphate buffered saline
Gene Alias arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein
Host Species Rabbit
Immunogen Synthetic peptide derived from human FLAP protein.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 241
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.