Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FLAP Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518283
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
ALOX5AP is a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
Specifications
FLAP | |
Polyclonal | |
Unconjugated | |
ALOX5AP | |
5-lipoxygenase-activating protein FLAP; Alox5ap; arachidonate 5-lipoxygenase activating protein; arachidonate 5-lipoxygenase-activating protein; Flap; MK-886-binding protein | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
241 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P20292 | |
ALOX5AP | |
Synthetic peptide sequence (KTISTTISPLLLIP) corresponding to the C-terminus amino acids of ALOX5AP (aa 148-161). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction