Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582926 Supplier Invitrogen™ Supplier No. PA582926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKVEHESRTQ NGRSFQRTGT LAFERVYTAN QNCVDAYPTF LAVL Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 89%.

ALOX5AP is a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLAP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALOX5AP
Gene Accession No. P20292
Gene Alias 5-lipoxygenase-activating protein FLAP; Alox5ap; arachidonate 5-lipoxygenase activating protein; arachidonate 5-lipoxygenase-activating protein; Flap; MK-886-binding protein
Gene Symbols ALOX5AP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALOX5AP. Recombinant protein control fragment (Product #RP-89567).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.