Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLASH Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14168

Catalog No. PA14168


Only null left
Add to Cart

Description

Description

PA1-4168 detects the FLASH protein in mouse samples. PA1-4168 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~212 kDa protein representing FLASH in NIH3T3 cells. The PA1-4168 immunogen is a synthetic peptide corresponding to residues W(1909) T R N D D R E I L L E(1920) of mouse FLASH.

This protein is highly similar to FLASH, a mouse apoptotic protein identified by its interaction with the death-effector domain (DED) of caspase 8. Studies of FLASH protein suggested that this protein may be a component of the death-inducing signaling complex that includes Fas receptor, Fas-binding adapter FADD, and caspase 8, and plays a regulatory role in Fas-mediated apoptosis. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FLASH
Polyclonal
Unconjugated
CASP8AP2
AA387232; CASP8 associated protein 2; CASP8AP2; CASP8-associated protein 2; caspase 8 associated protein 2; CED-4; D4Ertd659e; FLASH; FLASH homolog RIP25; FLICE associated huge; FLICE-associated huge protein; FLJ11208; human FLASH; KIAA1315; mKIAA1315; RIP25; RP11-63K6.3
Rabbit
Antigen affinity chromatography
RUO
26885
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9WUF3
CASP8AP2
Synthetic peptide corresponding to residues W(1909) T R N D D R E I L L E(1920) of mouse FLASH.
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.