Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ System DNA Cassettes

Catalog No. p-4684347
Click to view available options
Separation Range:
10 to 1000 bp
50 to 4000 bp
Wells:
12 + 1-well
16 + 1-well

Cassettes optimized for DNA migration. Lonza™ FlashGel™ System DNA Cassettes separate DNA quickly and DNA migration can be seen as it happens. Separates DNA in 2 to 7 minutes.

  • Get results in 5 minutes or less
  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2.2%
Product Type DNA Cassettes
Separation Range 10 to 1000 bp
Wells 12 + 1-well
Formulation 2.2% Agarose
For Use With (Application) Electrophoresis
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Storage Requirements 18° to 26°C
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.