Learn More
BUCHI FlashPure ID Silica Bundle
Shop All BUCHI Products
Click to view available options
:
FlashPure ID Silica Bundle
Description
- Integrated RFID technology automatically transfers cartridge settings and data to the flash system
- Silica HP provides higer performance for polar compounds using organic solvents
- RFID for optimal convenience on BUCHI Reveleris™ systems
- 40μm irregular particle
- Selection of cartridge sizes
Specifications
Specifications
| Material | Polyethylene |
| Weight (Metric) | 4 to 12 g |
| Type | HP Silica |
| For Use With (Application) | Difficult Separations, Polar Compounds |