Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Flat Head Axe with Fiberglass Handle
SDP

Catalog No. 19173546
Click to view available options
Weight (English):
10.10 lb. (8 lb. Head Size)
8.00 lb (6.00 lb. Head Size)

Non-sparking flat-head axe that is ideal for use in hazardous or high-risk environments.

  • Bronze head
  • Length: 36 in.

Specifications

Product Type Flat Head Axe
Length (English) Blade 5 in.
Weight (English) 8.00 lb (6.00 lb. Head Size)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.