Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%

Catalog No. aaa14495mc
Click to view available options
Quantity:
100 mg
500 mg

CAS: 1891059-93-0 | C27H31N9Na2O15P2 | 829.52 g/mol

Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

Solubility
Soluble in water (50 mg/ml).

Notes
Store at 4°C. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1891059-93-0
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
PubChem CID 131675332
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Quantity 100 mg
Solubility Information Soluble in water (50mg/ml).
Formula Weight 829.52
Percent Purity 94%
Assay (dry wt.), water <10%
Chemical Name or Material Flavin adenine dinucleotide disodium salt hydrate
EINECSNumber 282-733-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.