Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flavin containing monooxygenase 4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Flavin containing monooxygenase 4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Flavin containing monooxygenase 4 Polyclonal specifically detects Flavin containing monooxygenase 4 in Rat samples. It is validated for Western Blot.Specifications
Flavin containing monooxygenase 4 | |
Polyclonal | |
Rabbit | |
Q8K4B7 | |
2329 | |
Synthetic peptides corresponding to Fmo4 (flavin containing monooxygenase 4) The peptide sequence was selected from the middle region of Fmo4. Peptide sequence PGIHKFKGQILHSQEYRIPDAFRGKRILVVGLGNTGGDVAVELSGIAAQV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
dimethylaniline monooxygenase [N-oxide-forming] 4, Dimethylaniline oxidase 4, EC 1.14.13.8, flavin containing monooxygenase 4, FMO 4, FMO2, Hepatic flavin-containing monooxygenase 4 | |
FMO4 | |
IgG | |
64 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title