Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Flavin reductase Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537842
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).
Specifications
Flavin reductase | |
Polyclonal | |
Unconjugated | |
BLVRB | |
Biliverdin reductase B; biliverdin reductase B (flavin reductase (NADPH)); biliverdin-IX beta-reductase; BLVRB; BVRB; BVR-B; epididymis secretory protein Li 10; flavin reductase (NADPH); FLR; FR; GHBP; green heme-binding protein; HEL-S-10; NADPH; NADPH-dependent diaphorase; NADPH-flavin reductase; SDR43U1; short chain dehydrogenase/reductase family 43U, member 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
645 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P30043 | |
BLVRB | |
Peptide with sequence C-SRLPSEGPRPAH, from the internal region of BLVRB (aa38-49). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction